Order Entry
Ireland
ContactUsLinkComponent
 
Polysorbate 20, clear yellow liquid
  ICNA04806576
 :  
Polysorbate 20, clear yellow liquid
  ICNA04806576
 :  

 

  • Pk:
    100 g
  • Seq:
    3401

 

 

  • Identity Test A:
    Passes
  • Identity Test B:
    Passes
  • FTIR:
    Conforms to Reference
  • Solubility (1 g/20 ml):
    Clear, Foamy Solution
  • Specific Gravity:
    1.08 - 1.13
  • Water Content:
    ≤3%
  • Saponification Number:
    40 - 50

 

 

 :
 :Ambient
 :9005-64-5
 :C₅₈H₁₁₄O₂₆
 :MFCD00165986