Order Entry
Ireland
ContactUsLinkComponent
Anti-CD334 Mouse Monoclonal Antibody (PerCP) [clone: HIM3-4]
Anti-CD334 Mouse Monoclonal Antibody (PerCP) [clone: HIM3-4]
Catalog # ABNOMAB13878
Supplier:  Abnova
Anti-CD334 Mouse Monoclonal Antibody (PerCP) [clone: HIM3-4]
Catalog # ABNOMAB13878
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD334
  • Clonality:
    Monoclonal
  • Clone:
    HIM3-4
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Gene ID:
    945
  • Size:
    100 reactions
  • Pk:
    100 Assays

Specifications

About this item

Mouse monoclonal antibody raised against human CD33.

Type: Primary
Antigen: CD334
Clonality: Monoclonal
Clone: HIM3-4
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human