Order Entry
Ireland
ContactUsLinkComponent
 
Anti-MUC1 Mouse Monoclonal Antibody [clone: HMPV]
  ABNOMAB13356
 :  Abnova
Anti-MUC1 Mouse Monoclonal Antibody [clone: HMPV]
  ABNOMAB13356
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen symbol:
    MUC1
  • Clonality:
    Monoclonal
  • Clone:
    HMPV
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Gene ID:
    4582
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

Mouse monoclonal antibody raised against native human MUC1.

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: HMPV
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human