Order Entry
Ireland
ContactUsLinkComponent
Anti-Hspg2 Rat Monoclonal Antibody [clone: A7L6]
Anti-Hspg2 Rat Monoclonal Antibody [clone: A7L6]
Catalog # ABNOMAB13314
Supplier:  Abnova
Anti-Hspg2 Rat Monoclonal Antibody [clone: A7L6]
Catalog # ABNOMAB13314
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Hspg2
  • Clonality:
    Monoclonal
  • Clone:
    A7L6
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human,
    Mouse
  • Gene ID:
    15530
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Rat monoclonal antibody raised against native mouse Hspg2.

Type: Primary
Antigen: Hspg2
Clonality: Monoclonal
Clone: A7L6
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Human,Mouse