Order Entry
Ireland
ContactUsLinkComponent
 
Anti-Streptococcus group A Rabbit Polyclonal Antibody
Catalog # ABNOPAB29814
Supplier:  Abnova
Anti-Streptococcus group A Rabbit Polyclonal Antibody
Catalog # ABNOPAB29814
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Streptococcus group A antigen
  • Antigen symbol:
    Streptococcus group A
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Streptococcus
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Rabbit polyclonal antibody raised against Streptococcus group A.

Type: Primary
Antigen: Streptococcus group A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Streptococcus