Order Entry
Ireland
ContactUsLinkComponent
Anti-KRT19 Mouse Monoclonal Antibody (FITC) [clone: A53-B/A2]
Anti-KRT19 Mouse Monoclonal Antibody (FITC) [clone: A53-B/A2]
Catalog # ABNOMAB13967
Supplier:  Abnova
Anti-KRT19 Mouse Monoclonal Antibody (FITC) [clone: A53-B/A2]
Catalog # ABNOMAB13967
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    KRT19
  • Clonality:
    Monoclonal
  • Clone:
    A53-B/A2
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Gene ID:
    3880
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against native KRT19.

Type: Primary
Antigen: KRT19
Clonality: Monoclonal
Clone: A53-B/A2
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human