Order Entry
Ireland
ContactUsLinkComponent
Fast DNA™ SPIN kit for soil, MP Biomedicals
Fast DNA™ SPIN kit for soil, MP Biomedicals
Catalog # ICNA116560200
Supplier:  MP Biomedicals
CAS Number:  
undefined
Fast DNA™ SPIN kit for soil, MP Biomedicals
Catalog # ICNA116560200
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Soil
  • Target molecule:
    DNA
  • Description:
    Fast DNA™ SPIN Kit for soil, 50×2 ml preps
  • Description:
    Fast DNA™ SPIN Kit for soil

Specifications

About this item

The Fast DNA™ SPIN Kit for Soil is designed to efficiently isolate bacterial, fungal, plant and animal genomic DNA from soil and other environmental samples. It quickly and efficiently isolates PCR-ready genomic DNA directly from soil samples in less than 60 minutes.

  • Isolation of DNA from cells present in soil or other environmental samples
  • Rapid and reproducible sample lysis with the FastPrep®-24 or FastPrep® FP120 instrument
  • Easily isolate DNA from a variety of organisms in many different types of soil
  • Lyse and isolate DNA in less than 60 minutes
  • No hazardous organic reagents required

The FastDNA® SPIN Kit is designed for use with any FastPrep® instrument. Plant and animal tissues, bacteria, algae, fungi spores and other members of a soil population are easily lysed within 40 seconds. These benchtop devices use a unique, optimised motion to homogenise samples by simultaneous impaction with lysing matrix particles.

For research purposes only.

Delivery: Kit contains Lysing Matrix E, 50×2,0 ml tubes; Sodium Phosphate Buffer, 60 ml; MT Buffer, 8 ml; PPS Solution, 25 ml; Binding Matrix, 66 ml; SPIN Modules, 50 each; Catch Tubes, 50 each; Concentrated SEWS-M, 12 ml; DES, 20ml; and BBS Gel Loading Dye, 200 µl.