Oversized breaker lockout device
Supplier: Brady
Certificates
About this item
This oversized 480/600 V frame lockout fits breakers up to 57,15 mm wide and 22,2 mm thick.
- Made of durable PP and glass filled impact-modified nylon
- Applies in seconds with thumbscrew mechanism so no screwdriver is needed
- New blade design with improved grip works on even more breaker models
Frequently Bought Together