Visual colour kits, alkalinity, AL-AP
705-0135
: Hach
:
- Pk:1 KIT
- Description:Alkalinity (CaCO₃)
- Tests per kit:100
- Range:5 - 100 mg/l
Caused by the presence of carbonates, bicarbonates, hydroxides, and other dissolved salts, alkalinity is important to food and beverage processors, boiler operators, and aquatic biologists.
- Kit utilises drop count titration method
- Simple, economical titration method
: Rugged kit contains all required reagents, solutions, and glassware.