Ultra-low temperature freezers, upright models, −86 °C
Supplier: DAIREI EUROPE
Certificates
About this item
ULUF −86 °C range freezers are manufactured with the true and original single compressor technology.
- Optimised cooling system and lower energy consumption
- Castors and door key lock
- Insulated inner doors
- Filterless construction, which prevents problems due to dust buildup on the filter
- Micro processor controller available with every possible alarm feature, integrated data logger, contact for remote alarm, RS485/RS232 interface and USB read out
Specifications
- Catalog No:
- DAIRDAI1375
- DAIRDAI1416
- DAIRDAI1408
- Model:
- ULUF 550
- ULUF 750
- ULUF 450
- Cat. No.:
- DAIRDAI1375
- DAIRDAI1416
- DAIRDAI1408
- Electrical:
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz