Order Entry
Ireland
ContactUsLinkComponent
Gas detection tubes for phosgene
Gas detection tubes for phosgene
Supplier:  Dräger
Certificates
undefined
Gas detection tubes for phosgene
Supplier:  Dräger
Specifications
  • Catalog No:
  • DRAECH28301
  • 151-3148
  • Model:
  • Phosgene 0,25/c
  • Phosgene 0,02/a
  • Colour change:
  • Yellow to blue green
  • White to red
  • Humidity:
  • 3 - 15 mg H₂O/l
  • 1 - 18 mg H₂O/l
  • No. of strokes:
  • 5
  • 20/40
  • Standard deviation:
  • ±15 - 20 %
  • ±10 - 15 %
  • Standard range:
  • 0,25 - 5 ppm
  • 0,02 - 1 ppm/ 0,02 - 0,6 ppm