Order Entry
Ireland
ContactUsLinkComponent
Impact test according to CSA
Impact test according to CSA
Catalog # SIMESP1896
Supplier:  Industrial Physics
undefined
Impact test according to CSA
Catalog # SIMESP1896
Supplier:  Industrial Physics
Specifications
  • Description:
    Impact tester according to Canadian Standards Association (CSA)
  • Seq:
    04
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Impact test used to determine the impact resistivity and flexibility of materials.
  • Spirit level built-in for correct positioning
  • Dual scale instrument equipped with a special guidance
  • Distance between each impact is always according to the standards
Certifications: Conforms to the Canadian Standards Association (CSA).