Order Entry
Ireland
ContactUsLinkComponent
Chemical resistant cleanroom overalls, Kimtech™ A8
Chemical resistant cleanroom overalls, Kimtech™ A8
Supplier:  KIMBERLY CLARK
   Sustainable Options Available
Certificates
undefined
Chemical resistant cleanroom overalls, Kimtech™ A8
Supplier:  KIMBERLY CLARK
   Sustainable Options Available
Specifications
  • Catalog No:
  • 113-8622
  • 113-8627
  • 113-8625
  • 113-8621
  • 113-8626
  • 113-8623
  • 113-8624
  • Model:
  • A8
  • A8
  • A8
  • A8
  • A8
  • A8
  • A8
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Anti-static:
  • +
  • +
  • +
  • +
  • +
  • +
  • +
  • Cleanroom class:
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • Garment application:
  • Pharmaceutical, Laboratory
  • Pharmaceutical, Laboratory
  • Pharmaceutical, Laboratory
  • Pharmaceutical, Laboratory
  • Pharmaceutical, Laboratory
  • Pharmaceutical, Laboratory
  • Pharmaceutical, Laboratory
  • Bacterial filtration efficiency:
  • 94%
  • 94%
  • 94%
  • 94%
  • 94%
  • 94%
  • 94%
  • Particle shedding (Helmke Drum):
  • Level III
  • Level III
  • Level III
  • Level III
  • Level III
  • Level III
  • Level III
  • Weight:
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²