Order Entry
Ireland
ContactUsLinkComponent
 
Anti-CPB2 Rabbit Polyclonal Antibody
Catalog # BSBTPB9997
Supplier:  Boster Bio
undefined
Anti-CPB2 Rabbit Polyclonal Antibody
Catalog # BSBTPB9997
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    carboxypeptidase B2 (plasma)
  • Antigen symbol:
    CPB2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Gene ID:
    1361
  • Antigen synonyms:
    TAFI|CPU|PCPB
  • Storage temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    E. coli-derived human CPB2 recombinant protein (Position: K166-D388). Human CPB2 shares 86.1% and 85.7% amino acid (aa) sequence identity with mouse and rat CPB2, respectively.
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Carboxypeptidase B2 (CPB2), also known as carboxypeptidase U (CPU), plasma carboxypeptidase B (pCPB) or thrombin-activatable fibrinolysis inhibitor (TAFI), is an enzyme that, in humans, is encoded by the gene CPB2. CPB2 is synthesized by the liver and circulates in the plasma as a plasminogen-bound zymogen. When it is activated by proteolysis at residue Arg92 by the thrombin/thrombomodulin complex, CPB2 exhibits carboxypeptidase activity. Activated CPB2 reduces fibrinolysis by removing the fibrin C-terminal residues that are important for the binding and activation of plasminogen.

1. Boffa MB, Reid TS, Joo E, Nesheim ME, Koschinsky ML (May 1999). "Characterization of the gene encoding human TAFI (thrombin-activable fibrinolysis inhibitor; plasma procarboxypeptidase B)". Biochemistry 38 (20): 6547–58.
2. Eaton DL, Malloy BE, Tsai SP, Henzel W, Drayna D (Dec 1991). "Isolation, molecular cloning, and partial characterization of a novel carboxypeptidase B from human plasma". J Biol Chem 266 (32): 21833–8.
3. Tsai SP, Drayna D (Dec 1992). "The gene encoding human plasma carboxypeptidase B (CPB2) resides on chromosome 13". Genomics14 (2): 549–50.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.