Order Entry
Ireland
ContactUsLinkComponent
Beakers, aluminium
Beakers, aluminium
Catalog # 39047.KT
Supplier #: 39047.KT
Beakers, aluminium
Catalog # 39047.KT
Supplier #: 39047.KT
Log in for availability
Select an Option
63 mm
Select an Option
250 ml
Select an Option
92 mm
Select an Option
01
Select an Option
1
About this item
Beaker, aluminium, with pourout lip.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Aluminum is the most abundant metallic element on the earth and has many desirable physical and chemical properties. A highly impervious oxide film (approximately 5 nm thick) is resistant to corrosion by seawater, and other aqueous and chemical solutions. Additionally, this element offers chemical stability in the presence of most organic compounds. Aluminum's reactivity increases with temperature, therefore, the use of aluminum labware at high temperatures is not recommended.
Frequently Bought Together