Order Entry
Ireland
ContactUsLinkComponent
DAB substrate
DAB substrate
Catalog # PIER34002
CAS Number:  
undefined
DAB substrate
Catalog # PIER34002
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    DAB Substrate Kit, sufficient for 25 uses at 10 ml each
  • Seq:
    01

Specifications

About this item

DAB (3,3'-diaminodbenzidine) is a sensitive colorimetric substrate used with horseradish peroxidase (HRP) in immunohistochemistry and immunoblotting applications. HRP catalyses hydrogen peroxide oxidation of substrates such as DAB. Electrons are transferred by HRP from the DAB to the peroxide to yield an insoluble brown product.

Delivery: Kit contains DAB Solution (10X), 25 ml; Stable Peroxide Substrate Buffer, 250 ml.