Order Entry
Ireland
ContactUsLinkComponent
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride Pd 15,8 --> 16,8 %
Irritant
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride Pd 15,8 --> 16,8 %
Certificates
undefined
Irritant
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride Pd 15,8 --> 16,8 %
Specification Test Results
  • Appearance:
    Beige to dark brown Powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    10.1 to 11.6 % (Cl)
  • Palladium (Pd):
    15.8 to 16.8 %