
2-Methylfuran-3-boronic acid pinacol ester 90%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:864776-02-3
:
Formula:C₁₁H₁₇BO₃
MDL Number:MFCD09879905
ADR:6.1,III
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to orange Liquid
- Infrared spectrum:Conforms
- GC:≥89.0 %
Specification Test Results
Frequently Bought Together