6-O-Palmitoyl-L-ascorbic acid, meets analytical specification of Ph. Eur., USP
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: L-Ascorbic acid 6-palmitate, Palmitoyl-L-ascorbic acid, Ascorbyl Palmitate, (S)-2-((R)-3,4-Dihydroxy-5-oxo-2,5-dihydrofuran-2-yl)-2-hydroxyethyl palmitate, L(+)-Ascorbyl palmitate
CAS Number:137-66-6
:
Formula:C₂₂H₃₈O₇
MDL Number:MFCD00005377
:414.54 g/mol
Boiling Pt:250 °C
Melting Pt:111…115 °C
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Melting point:107 to 117 °C
- Titration Iodimetric:98.0 to 100.5 % (on dry substance)
- Loss on drying:≤1 % (60 °C, 5 Hrs) (vacuum)
- Heavy metals:≤10 ppm
- Appearance of solution:Passes test
- Residue after ignition:≤0.1 %
- Specific optical rotation:+21° to +24° (20 °C, 589 nm) (c=10, methanol)
Specification Test Results
Frequently Bought Together