Order Entry
Ireland
ContactUsLinkComponent
Mercury(II) bromide ACS
Mercury(II) bromide ACS
Catalog # ACRO448121000
CAS Number:  
undefined
Mercury(II) bromide ACS
Catalog # ACRO448121000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to very light yellow Powder
  • Titration Complexometric:
    ≥98.0 %
  • Chloride (Cl):
    ≤0.25 %
  • Insoluble matter:
    ≤0.05 % (In methanol)
  • Residue after reduction:
    ≤0.02 %

Specification Test Results

About this item

Synonyms: Mercuric bromide, Mercury dibromide
 :
 :360.4 g/mol
Boiling Pt:322 °C (760 mmHg)
Melting Pt:236 °C
Density:6,1 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1634
ADR:6.1,II
 :GHS06
 :GHS08
 :GHS09
CAS Number:7789-47-1
Formula:Br₂Hg
MDL Number:MFCD00011040