Order Entry
Ireland
ContactUsLinkComponent
 
 
 
 
Ammonium metavanadate ACS
 
undefined
 
 
 
Ammonium metavanadate ACS
 

 

  • Appearance:
    White to yellow to tan Powder
  • Solubility:
    (in Ammonium hydroxide) Passes test
  • Titration with KMnO4:
    ≥99.0 %
  • Chloride (Cl):
    ≤0.2 %
  • Carbonate (CO3):
    (Limit about 0.3 %) Passes test
  • Sulfate (SO4):
    ≤0.05 %

 

 

Frequently Bought Together