
4-Fluoro-3-(tetrazol-5-yl)phenylboronic acid 95%
Catalog # ACRO445460010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to off-white Solid
- Infrared spectrum:Conforms
- Additional info:May contain varying amounts of anhydrides
- HPLC:≥94.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:207.96 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1009303-56-3
Formula:C₇H₆BFN₄O₂
MDL Number:MFCD13176533