


Ethanol denatured 96%, Technical Grade (denatured with 5% methanol)
ACRO444140025
:
- Pack type:Plastic bottle
- Pk:2,5 L
- Seq:5096
- Appearance:Clear colorless Liquid
- Refractive index:1.3600 to 1.3630 (20°C, 589 nm)
- Density:(20°C) 0.780 to 0.807 kg/l
- GC:≤5 % (methanol)
:
:46.07 g/mol
:∼78 °C (1013 hPa)
:–117 °C
:0,8 g/cm³
:12 °C
:1170
:3,II
:GHS02
:GHS07
:GHS08
:Ambient
:64-17-5
:H₃CCH₂OH
:MFCD00003568