

Methyl ethyl ketone, Technical Grade
Catalog # ACRO444170050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Metal drum
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.2 %
- GC:≥99.0 %
- Color scale:≤10 APHA
Specification Test Results
About this item
:
Boiling Pt:80 °C (760 mmHg)
Melting Pt:–87 °C
Density:0,806 g/cm³
Flash Pt:–7 °C
Storage Temperature:Ambient
UN:1193
ADR:3,II
:72.11 g/mol
:GHS07
:GHS02
CAS Number:78-93-3
Formula:C₂H₅COCH₃
MDL Number:MFCD00011648