Chlorobenzene, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-90-7
:
Formula:C₆H₅Cl
MDL Number:MFCD00000530
:112.56 g/mol
Boiling Pt:131 °C (760 mmHg)
Melting Pt:–45 °C
Density:1,1075 g/cm³
Flash Pt:23 °C
UN:1134
ADR:3,III
:GHS07
:GHS09
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.4 %
Specification Test Results
Related Information
Frequently Bought Together