(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide 98%
Catalog # ACRO439341000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Yellow to orange Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Specific optical rotation:+54.0° to +58.0° (20°C, 589 nm) (c=1, acetone)
Specification Test Results
About this item
:
:272.192 g/mol
Melting Pt:227,7…229.4 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:95713-52-3
Formula:C₉H₉FN₄O₅
MDL Number:MFCD00042049