2,4,6-Trimethylstyrene 95%
Catalog # ACRO436280050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥94.0 %
- Stabilizer:≤0.05 % tert-Butylcatechol
Specification Test Results
About this item
:
:146.23 g/mol
Boiling Pt:209 °C
Density:0,906 g/cm³
Flash Pt:75 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:769-25-5
Formula:C₁₁H₁₄
MDL Number:MFCD00008613