2,4,6-Trifluorophenylboronic acid 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:182482-25-3
:
Formula:C₆H₄BF₃O₂
MDL Number:MFCD01863169
Melting Pt:239,5…247.5 °C
:175.9 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥94.0 %
Specification Test Results
Frequently Bought Together