7-Amino-1-tetralone 97%
:22009-40-1
:
:C₁₀H₁₁NO
:MFCD07369817
:161.2 g/mol
:138…140 °C
:GHS07
:Ambient
- Appearance:Yellow to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:135 to 140 °C
- GC:≥96.0 %
- Residue after evaporation:≤0.001 %
Frequently Bought Together