
4,4,5,5-Tetramethyl-2-(tetrahydrofuran-3-yl)-1,3,2-dioxaborolane 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:331958-90-8
:
Formula:C₁₀H₁₉BO₃
MDL Number:MFCD09878900
:198.069 g/mol
Flash Pt:93 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together