

Anisole, extra dry 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-66-3
:
Formula:C₇H₈O
MDL Number:MFCD00008354
:108.14 g/mol
Boiling Pt:154 °C (760 mmHg)
Melting Pt:–37 °C
Density:0,99 g/cm³
Flash Pt:45 °C
Storage Temperature:Ambient
UN:2222
ADR:3,III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥98.5 %
- Residue after evaporation:≤0.005 %
- UV:at 200 nm A: ≤0.75
- Non-volatile matter:≤0.0003 %
- Free acid (CH3COOH):≤0.0005 %
Specification Test Results
Related Information
Frequently Bought Together