Chitosan, M.W. 600,000-800,000
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9012-76-4
:MFCD00149018
MDL Number:MFCD00161512
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Off-white to beige Powder
- Infrared spectrum:Conforms
- Assay:≥90 % (DAC)
- Loss on drying:≤10.0 %
- Molecular weight:600000 to 800000 Da
- Viscosity:50 to 200 mPa.s
- Ash:≤1.5 %
Specification Test Results
Frequently Bought Together