Order Entry
Ireland
ContactUsLinkComponent
Methyl tricosanoate 99+%
Corrosive
Methyl tricosanoate 99+%
Certificates
undefined
Corrosive
Methyl tricosanoate 99+%
Certificates

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    53 to 56 °C
  • GC:
    ≥99.0 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

Related Information

Frequently Bought Together