Diammonium hydrogen citrate 98%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3012-65-5
:
Formula:HOC(CO₂H)(CH₂CO₂NH₄)₂
MDL Number:MFCD00013068
:226.19 g/mol
Density:1,5 g/cm³
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Titration with HClO4:97.5 to 102.5 %
Specification Test Results
Frequently Bought Together