Order Entry
Ireland
ContactUsLinkComponent
D-(+)-Glucose, anhydrous Ph. Eur., USP, BP
D-(+)-Glucose, anhydrous Ph. Eur., USP, BP
Certificates
undefined
D-(+)-Glucose, anhydrous Ph. Eur., USP, BP

Specification Test Results

  • Appearance:
    White to almost white Crystalline powder
  • Assay:
    97.5 to 102.0 % (Anhydrous substance)
  • Water:
    ≤1.0 % (K.F.)
  • Loss on drying:
    ≤0.5 % (1 g, 105°C, 16 h)
  • Sulfated ash:
    ≤0.1 % (1 g, 600°C)
  • Appearance of solution:
    (EP) Passes test
  • Identification:
    Passes test (EP, USP)
  • Related substances:
    ≤0.4 % (sum of impurities A and B)
  • Specific optical rotation:
    +52.5° to +53.3° (20°C, 589 nm) (C=10,H2O/dilute NH3) (Anhydrous substance)
  • Acidity:
    (USP) Passes test
  • Color of solution:
    (USP) Passes test
  • Chloride (Cl):
    ≤0.018 %
  • Sulfate (SO4):
    ≤0.025 %
  • Arsenic (As):
    ≤1 µg/g
  • Soluble starch, sulfite:
    ≤15 ppm (EP)
  • Dextrin:
    (EP, USP) Passes test
  • Conductivity:
    ≤20 µS/cm

Specification Test Results

Related Information

Frequently Bought Together