Order Entry
Ireland
ContactUsLinkComponent
Citric acid monohydrate 99+% ACS
Irritant
Citric acid monohydrate 99+% ACS
Certificates
undefined
Irritant
Citric acid monohydrate 99+% ACS

Specification Test Results

  • Appearance:
    Colorless to white Powder or crystals
  • Residue after ignition:
    ≤0.02 %
  • Titration with NaOH:
    99.0 to 102.0 %
  • Carboniz. sub. by hot H2SO4:
    Passes test
  • Chloride (Cl):
    ≤0.001 %
  • Oxalate (C2O4):
    (limit about 0.05%) Passes test
  • Phosphate (PO4):
    ≤0.001 %
  • Sulfate (SO4):
    ≤0.002 %
  • Iron (Fe):
    ≤3 ppm
  • Lead (Pb):
    ≤2 ppm
  • Insoluble matter:
    ≤0.005 %

Specification Test Results

Related Information

Frequently Bought Together