
10-Bromo-1-decanol 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:53463-68-6
:
Formula:HO(CH₂)₉CH₂Br
MDL Number:MFCD00041681
:237.18 g/mol
Boiling Pt:165…169 °C (10 mmHg)
Density:1,19 g/cm³
Flash Pt:109 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4780 to 1.4800 (20°C, 589 nm)
- GC:≥94 %
Specification Test Results
Frequently Bought Together