

Tetraethylammonium perchlorate 0.2M in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2567-83-1
:
Formula:C₈H₂₀ClNO₄
MDL Number:MFCD00041977
:229.7 g/mol
Melting Pt:> 300 °C
Density:1,0016 g/cm³
Storage Temperature:Ambient
UN:3139
ADR:5.1,II
:GHS03
:GHS07
Specification Test Results
- Appearance:Clear colorless Solution
- Strength:0.2 M Exact strength on the certificate of analysis
Specification Test Results
Frequently Bought Together