Epiandrosterone 99%
Catalog # ACRO215390050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:171 to 177 °C
- Loss on drying:≤0.5 %
- GC:≥98.5 %
- Specific optical rotation:+88° to +94° (20 °C, 589 nm) (c=1, CH3OH)
Specification Test Results
About this item
:
:290.447 g/mol
Melting Pt:171…177 °C
Storage Temperature:Ambient
CAS Number:481-29-8
Formula:C₁₉H₃₀O₂
MDL Number:MFCD00064134