Mercury(II) iodide, red ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7774-29-0
:
Formula:HgI₂
MDL Number:MFCD00011044
:454.4 g/mol
Boiling Pt:354 °C (760 mmHg)
Melting Pt:259 °C
Density:6,36 g/cm³ (20 °C)
UN:1638
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Red Crystalline powder
- Titration Iodometric:≥99.0 % (Dried basis)
- Solubility:(In potassium iodide soln.) Passes test
- Mercurous Mercury (as Hg):≤0.1 %
- Soluble Hg salts (as Hg):≤0.05 %
Specification Test Results
Frequently Bought Together