Horse Cytochrome C (from Heart muscle)
Catalog # ACRO147530010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Source:Heart muscle
- Species:Horse
- Storage conditions:Freezer
- MDL No.:MFCD00130890
- Protein/peptide name:Cytochrome C
- Purity:0.9
Specifications
About this item
- Red-brown to dark brown Powder