
2-Phenylpyridine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1008-89-5
:
Formula:C₁₁H₉N
MDL Number:MFCD00006280
:155.2 g/mol
Boiling Pt:268…270 °C (760 mmHg)
Melting Pt:76 - 77 °C
Density:1,086 g/cm³
Flash Pt:110 °C
ADR:6.1,III
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6220 to 1.6260 (20°C, 589 nm)
- GC:≥96.0 %
- Residue after evaporation:≤0.001 %
Specification Test Results
Frequently Bought Together