Sodium sulphate decahydrate, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7727-73-3
:
Formula:Na₂SO₄·10H₂O
MDL Number:MFCD00149189
:322.2 g/mol
Boiling Pt:1429 °C (1013 hPa)
Melting Pt:884 °C
Density:1,464 g/cm³ (20 °C)
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- pH:5.2 to 9.2 (5% solution)
- Titration after Ion exchange:≥99.0 %
- Chloride (Cl):≤0.02 %
- Calcium (Ca):≤0.01 %
- Iron (Fe):≤0.005 %
- Potassium (K):≤0.01 %
Specification Test Results
Frequently Bought Together