Order Entry
Ireland
ContactUsLinkComponent
β-Nicotinamide adenine dinucleotide hydrate 98+%
β-Nicotinamide adenine dinucleotide hydrate 98+%
Certificates
undefined
β-Nicotinamide adenine dinucleotide hydrate 98+%
Certificates

Specification Test Results

  • Appearance:
    White to yellow Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤11 % (K.F.)
  • Solubility:
    (1 % in water) Clear colorless
  • UV spectrophotometry:
    ≥98.0 %
  • Molar absorption coefficient:
    ≥17000 l/(mol.cm) (Lambda max 260 nm, pH 7.5)

Specification Test Results

Related Information

Frequently Bought Together