
Methyl-4-chloro-4-oxobutanoate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1490-25-1
:
Formula:C₅H₇ClO₃
MDL Number:MFCD00000750
:150.56 g/mol
Boiling Pt:58…65 °C (3 mmHg)
Density:1,232 g/cm³
Flash Pt:73 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4390 to 1.4410 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together