ESD safety ankle boots, lace-up, Miss L10, Sensation Lady Mid
: ELTEN
Casual, vintage style ESD ankle boot for ladies made from hydrophobised textile material in 'denim look', and hydrophobised nubuck leather, a padded tongue plus a low profile dual-density TPU/PU sole (Miss L10).
- Bioactive fabric lining
- Replaceable insole; orthopaedic insoles possible
- Full length inlay sole (ESD Pro Lady), grey
- Soft fleece ESD insole
- Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected. The shoe is tailored to the ergonomics of female feet.
Colour: Black/grey
: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC
- Catalog No:
- 114-8788
- 114-8787
- 114-8786
- 114-8785
- 114-8784
- 114-8783
- 114-8782
- 114-8781
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
Frequently Bought Together