Transport swabs for viruses and Chlamydia Sterilin
: STERILIN
These swabs are specially suited for the collection and transport of viruses and Chlamydia.
- Inert non-toxic PU sponge contains the liquid transport medium
- Swab tip is bathed in media at all times
- Contains special media for the optimal recovery of viral pathogens or Chlamydia
Suitable for in vitro use only.
: CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC.
: Individually wrapped in a unique plastic film pouch with an outer nitrogen flushed aluminium.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together