
1,3,2-Dioxathiolane-2,2-dioxide 96%
:1072-53-3
:
:C₂H₄O₄S
:MFCD00221769
:95…97 °C
:124.11 g/mol
:GHS07
:Refrigerator
- Appearance:White to beige Powder
- Infrared spectrum:Conforms
- Melting point:95 to 99 °C
- GC:≥95.0 %
Frequently Bought Together