Order Entry
Ireland
ContactUsLinkComponent
 
cis-5-Norbornene exo-2,3-dicarboxylic anhydride 95%
 
undefined
cis-5-Norbornene exo-2,3-dicarboxylic anhydride 95%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to tan Solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    140 to 145 °C
  • GC:
    ≥94 %

 

 

Frequently Bought Together