
1H-Pyrazole-4-carboxylic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:37718-11-9
:
Formula:C₄H₄N₂O₂
MDL Number:MFCD00011558
:112.09 g/mol
Melting Pt:282 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white Powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together